Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558135 Supplier Invitrogen™ Supplier No. PA558135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEFQPGSIRL EKHPKLPATQ PIPRIKSPNE MVGQKPLGEI TTVLGSHLIY KKRISNPFQG LSHRGSTISK GHKIQKTSDL KPSQTGPKEK Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 53%.

STOX1 (Storkhead box 1), also known as PEE4 or C10orf24, is a winged-helix domain containing transcription factor believed to play a role in the differentiation of stem cells. As a DNA-binding protein, STOX1 exists as three alternatively spliced isoforms, designated A, B and C, all of which localize to either the cytoplasm or the nucleus. Defects in STOX1 are the cause of pre-eclampsia/eclampsia 4 (PEE4), also known as gestational proteinuric hypertension. STOX1 was also found to be functionally involved in late-onset Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOX1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STOX1
Gene Accession No. Q6ZVD7
Gene Alias C10orf24; PEE4; storkhead box 1; storkhead-box protein 1; STOX1; Winged-helix domain-containing protein
Gene Symbols STOX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STOX1. Recombinant protein control fragment (Product #RP-97144).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.