Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562041 Supplier Invitrogen™ Supplier No. PA562041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLDERIPDRS QCTSPQPGTI TPSASGCVRE RTLPRNHCDS CHCCREDVHS THAPTLQRKS AKDCKDPYCP PSLCQVPPTE KSKSTVNFSY KTETLSKPK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.

The Storkhead box protein 2 (STOX2) is the only known paralog to STOX1, a winged-helix domain containing transcription factor believed to play a role in the differentiation of stem cells. STOX2 has been suggested to be part of a molecular profile unique to stem cells, and its mRNA may be part of a transcriptional profile observed with increased inflammatory response to air pollutants. Decreased STOX2 expression levels in decidua are also correlated with preeclampsia, suggesting STOX2 may play a role in the pathophysiology of preeclampsia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOX2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STOX2
Gene Accession No. Q9P2F5
Gene Alias 4933409N07Rik; AI449080; AI835114; KIAA1392; knockout CG10573-PA related; mKIAA1392; spinelinin; storkhead box 2; storkhead-box protein 2; Stox2
Gene Symbols STOX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STOX2. Recombinant protein control fragment (Product #RP-100223).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.