Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STPG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549489
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549489 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549489 Supplier Invitrogen™ Supplier No. PA549489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STPG1 Polyclonal Antibody for Western Blot

May positively contribute to the induction of apoptosis triggered by O(6)-methylguanine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STPG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STPG1
Gene Accession No. Q5TH74
Gene Alias 4930403G18Rik; 4930555I21Rik; C1orf201; MAPO2; O(6)-methylguanine-induced apoptosis 2; O6-methylguanine-induced apoptosis 2; sperm tail PG rich repeat containing 1; sperm tail PG-rich repeat containing 1; sperm-tail PG-rich repeat containing 1; sperm-tail PG-rich repeat-containing protein 1; STPG1; UPF0490 protein C1orf201; UPF0490 protein C1orf201 homolog
Gene Symbols STPG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 37-70 amino acids from the N-terminal region of human C1orf201.
Purification Method Antigen Affinity Chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90529
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.