Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STPG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144879
Description
Immunogen sequence: ATGSNAPFLS LTARESTFTI ASSIEKAVPG PGHYNVSEAQ KISRSPTLTR SVDAPSIPSC GKSYGYHIND DGSIIKCFPP ACD Antigen sequence identity: Mouse 37%; Rat 37%.
STPG2 (Sperm-tail PG-rich repeat-containing protein 2) associated diseases include Complex Chromosomal Rearrangement and Cone-Rod Dystrophy 18. An important paralog of this gene is STPG1.
Specifications
STPG2 | |
Polyclonal | |
Unconjugated | |
STPG2 | |
C4orf37; sperm tail PG-rich repeat containing 2; sperm-tail PG-rich repeat containing 2; sperm-tail PG-rich repeat-containing protein 2; STPG2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
285555 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N412 | |
STPG2 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 35-117. Recombinant protein control fragment (Product #RP-110146). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction