Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STPG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113245
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113245 Supplier Invitrogen™ Supplier No. PA5113245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STPG2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

STPG2 (Sperm-tail PG-rich repeat-containing protein 2) associated diseases include Complex Chromosomal Rearrangement and Cone-Rod Dystrophy 18. An important paralog of this gene is STPG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STPG2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene STPG2
Gene Accession No. Q8N412
Gene Alias C4orf37; sperm tail PG-rich repeat containing 2; sperm-tail PG-rich repeat containing 2; sperm-tail PG-rich repeat-containing protein 2; STPG2
Gene Symbols STPG2
Host Species Rabbit
Immunogen Recombinant Human Sperm-tail PG-rich repeat-containing protein 2 protein (201-459AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285555
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.