Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STPG4 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306297IR
Description
STPG4 Polyclonal specifically detects STPG4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
STPG4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
C2orf61 chromosome 2 open reading frame 61 | |
Produced in rabbits immunized with E. coli-derived Human STPG4 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
285051 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction