Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STPG4 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306297MFV450
Description
STPG4 Polyclonal antibody specifically detects STPG4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| STPG4 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| C2orf61 chromosome 2 open reading frame 61 | |
| Produced in rabbits immunized with E. coli-derived Human STPG4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 285051 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction