Learn More
STRA6 Monoclonal Antibody (496613)

Description
STRA6 Monoclonal Antibody for ICC/IF, Flow
Specifications
Specifications
| Antigen | STRA6 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 496613 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Stra6 |
| Gene Accession No. | Q9BX79 |
| Gene Alias | AI891933; FLJ12541; MCOPCB8; MCOPS9; PP14296; Receptor for retinol uptake STRA6; Retinoic acid-responsive protein; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; RGD1307551; stimulated by retinoic acid 6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6; stimulated by retinoic acid gene 6 homolog; stimulated by retinoic acid gene 6 protein; Stimulated by retinoic acid gene 6 protein homolog; STRA6; UNQ3126/PRO10282/PRO19578 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.