Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142887
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142887 Supplier Invitrogen™ Supplier No. PA5142887
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

STRA6 is a membrane protein involved in the metabolism of retinol. The encoded protein acts as a receptor for retinol/retinol binding protein complexes. This protein removes the retinol from the complex and transports it across the cell membrane. Defects in this gene are a cause of syndromic microphthalmia type 9 (MCOPS9).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRA6
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Stra6
Gene Accession No. O70491
Gene Alias AI891933; FLJ12541; MCOPCB8; MCOPS9; PP14296; Receptor for retinol uptake STRA6; Retinoic acid-responsive protein; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; RGD1307551; stimulated by retinoic acid 6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6; stimulated by retinoic acid gene 6 homolog; stimulated by retinoic acid gene 6 protein; Stimulated by retinoic acid gene 6 protein homolog; STRA6; UNQ3126/PRO10282/PRO19578
Gene Symbols Stra6
Host Species Goat
Immunogen Peptide with sequence QVWQKAECPQDPK.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20897
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.