Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRA6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2166 Supplier Bioss Supplier No. BS12351R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STRA6 is a membrane protein involved in the metabolism of retinol. The encoded protein acts as a receptor for retinol/retinol binding protein complexes. This protein removes the retinol from the complex and transports it across the cell membrane. Defects in this gene are a cause of syndromic microphthalmia type 9 (MCOPS9).

Specifications

Antigen STRA6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Stra6
Gene Accession No. Q9BX79
Gene Alias AI891933; FLJ12541; MCOPCB8; MCOPS9; PP14296; Receptor for retinol uptake STRA6; Retinoic acid-responsive protein; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; RGD1307551; stimulated by retinoic acid 6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6; stimulated by retinoic acid gene 6 homolog; Stimulated by retinoic acid gene 6 protein; Stimulated by retinoic acid gene 6 protein homolog; Stra6; UNQ3126/PRO10282/PRO19578
Gene Symbols Stra6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human STRA6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64220
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.