Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRA6, Rabbit anti-Mouse, Polyclonal, Millipore Sigma™

Catalog No. ABN166225UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN166225UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN166225UL Supplier MilliporeSigma Supplier No. ABN166225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-STRA6, Cat. No. ABN1662, is a highly specific rabbit polyclonal antibody that targets Stimulated by retinoic acid gene 6 protein (STRA6) and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Stimulated by retinoic acid gene 6 protein (UniProt: O70491; also known as Retinoic acid-responsive protein, STRA6) is encoded by the Stra6 gene (Gene ID: 20897) in murine species. STRA6 is a multi-pass membrane protein that acts as a high-affinity cell-surface receptor for the complex retinol-retinol binding protein (RBP/RBP4). It has one intramembrane and nine transmembrane helices in a dimeric assembly. It acts by removing retinol from RBP/RBP4 and transports it across the plasma membrane, where it can be metabolized. This mechanism does not depend on endocytosis. Binds to RBP/RBP4 with high affinity. Increases cellular retinol uptake from the retinol-RBP complex. STRA6 is widely expressed in the embryo and it in adults it is expressed choroid plexus and the brain microvascular cells that form the blood-organ barrier. It is also expressed in the retinal pigment epithelium, seminiferous epithelium in testis, the yolk sac, and in chorioallantoic placenta. Its expression in Sertoli cells depends on the stage of the spermatogenic cycle. STRA6 can be induced by retinoic acid and it is shown to be synergistically up-regulated by Wnt1 and retinoid in mammary epithelial cells. Mutations in Stra6 gene have been linked to the Matthew-Wood syndrome that is characterized by pulmonary agenesis/hypoplasia; microphthalmia/anophthalmia; and congenital cardiac, digestive, and urogenital malformations. (Ref.: Sadowski, S et al. (2017). Birth Defects Res. 109(4); 251-253; Chen, Y et al. (2016). Science 353 (6302), aad8266).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRA6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation Analysis: A representative lot detected STRA6 in 293T cells transfected with an empty vector or pCMVTag2b-Stra6 ( Courtesy of Dr. Noa Noy ). Western Blotting Analysis: A representative lot detected STRA6 in Western Blotting applications (Berry, D.C., et. al. (2012). Mol Cell Biol. 32(19):3851-9). Western Blotting Analysis: A representative lot detected STRA6 in 293T cells transfected with an empty vector or pCMVTag2b-Stra6 ( Courtesy of Dr. Noa Noy ). Flow Cytometry Analysis: A 1:100 dilution from a representative lot detected STRA6 in HT-29 cells. Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected STRA6 in mouse testis and human liver cancer tissues.
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Stimulated by retinoic acid gene 6 protein;Retinoic acid-responsive protein
Gene Symbols Stra6
Host Species Rabbit
Immunogen His-tggged recombinant fragment corresponding to 109 amino acids from the C-terminal region of murine Stimulated by retinoic acid gene 6 (STRA6) protein .
Molecular Weight of Antigen ∽90 kDa observed; 73.77 kda calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects murine Stimulated by retinoic acid gene 6 protein (STRA6). it targets an epitope within 109 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.