Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stra6 Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209269
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9269 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9269 Supplier Fabgennix Inc Supplier No. STRA6101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stra6 Polyclonal antibody specifically detects Stra6 in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Stra6
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Stra6
Gene Accession No. O70491, Q4QR83
Gene Alias AI891933; FLJ12541; MCOPCB8; MCOPS9; PP14296; Receptor for retinol uptake STRA6; Retinoic acid-responsive protein; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; RGD1307551; stimulated by retinoic acid 6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6; stimulated by retinoic acid gene 6 homolog; stimulated by retinoic acid gene 6 protein; Stimulated by retinoic acid gene 6 protein homolog; STRA6; UNQ3126/PRO10282/PRO19578
Host Species Rabbit
Immunogen Synthetic peptide taken within third large intra cytoplasmic loop within amino acid region 200-300 on rat Stra6 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20897, 363071
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.