Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stra8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984278
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4278 Supplier Bioss Supplier No. BS1903R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.

Specifications

Antigen Stra8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STRA8
Gene Accession No. Q7Z7C7
Gene Alias RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; Stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; STRA8
Gene Symbols STRA8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Stra8 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.