Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Stra8, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1656MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1656MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1656MI Supplier MilliporeSigma™ Supplier No. ABN1656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stimulated by retinoic acid gene 8 protein (UniProt P70278) is encoded by the Stra8 gene (Gene ID 20899) in murine species. Stra8 is an early retinoic acid (RA) responsive gene that encodes a cytoplasmic protein expressed exclusively in gonads, where it plays a pivotal role in initiating meiosis of both female and male germ cells. In female mice, Stra8 is expressed in embryonic gonocytes during a restricted time window and is indispensable for their entry into the meiotic S phase. In males, Stra8 expression is first detected in primitive spermatogonia at P5, then becomes prominent in premeiotic (preleptotene) spermatocytes from P10 onwards. Spermatogenesis, the transformation of stem cells into millions of haploid spermatozoa, is a highly regulated process that allows a constant spermatozoa production. Undifferentiated mitotic spermatogonia first develop into committed spermatogonia to produce spermatocytes, which then undergo meiosis to form haploid spermatids. Stra8 is not only required for meiosis, but also promotes spermatogonial differentiation. In the absence of Stra8, spermatogonial differentiation is impaired, giving rise to massive accumulations of undifferentiated and early differentiating spermatogonia (Type A spermatogonia) in aged testes. Stra8-deficient germ cells in postnatal male and fetal female mice arrest just before meiosis, without entering meiotic prophase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stra8
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% Sodium Azide.
Gene Stra8
Gene Accession No. P70278
Host Species Rabbit
Immunogen His-tagged recombinant full-length mouse Stra8.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_033318
Test Specificity This polyclonal antibody detected Stra8 immunoreactivity in testes from wild-type, but not Stra8-knockout mice (Mark, M., et al. (2008). J. Cell Sci. 121(Pt 19):3233-3242).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.