Learn More
Eisco™ Straight Adapters with Cone and Screw thread
SCI_ED
Catalog No. S29464 Shop All Eisco Products
Change view
Click to view available options
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
S29464 14/23
S29465 19/26
S29466 24/29
S29467 29/32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S29464 Supplier Eisco™ Supplier No. CH0835A
Only null left
Add to Cart
Add to Cart
Made from high quality borosilicate 3.3 glass and feature ground cone joints and plastic threaded screw caps with thread size 13 and 6-7 mm centered holes
  • Straight adapters
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
  • Specifications
    For Use With (Application) Connecting glassware
    Height (Metric) 8 cm
    Length (Metric) 1.7 cm
    Material Borosilicate Glass (3.3)
    Product Type Straight Cone Adapter
    Quantity 1
    Standard Taper Joint No. 14/23
    Width (Metric) 1.7 cm
    1 year limited manufacturer warranty