Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Straight Dissecting Scissors
SCI_ED

Catalog No. S17314 Shop All Medco Instruments Products
Click to view available options
Length (English):
4 in.
This item is not returnable. View return policy

Catalog No. S17314

Supplier: Medco Instruments 16MDCH

This item is not returnable. View return policy

Excellent for dissecting work.

Sharp points on finely finished, narrow blades make these chrome-plated steel scissors perfect for dissecting work. Length: 10cm.

Specifications

Blade Style Straight
Length (English) 4 in.
Length (Metric) 10 cm
For Use With (Application) Dissecting
Tip Style Sharp/Sharp
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.