Learn More
Dynalon™ Straight-Side PETE Containers
SCI_ED
For storing pastes, samples, powders, specimens etc. in various lab applications. Dynalon™ Straight-Side PETE Containers may be used below freezing to a maximum of 70°C.
Specifications
Material PETE
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S37265
SCI_ED
View Documents
Dynalon™
4263150032
1000 mL N/A N/A N/A
Description
Description
  • Containers are made of shatterproof, durable PETE with better gas barrier properties with H20, O2 and CO2
  • Transparent with white polypropylene liner less screw closures
Specifications
Specifications
PETE
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.