Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Straight/SPME Inlet Liner
SDP

Catalog No. 06711435 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-711-435 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-711-435 Supplier Restek™ Supplier No. 21112

missing translation for 'validMainframeMsg'

For Bruker/Varian GCs equipped with 1075/1077 inlets.

  • Geometry: Straight/SPME
  • Material: Borosilicate Glass
  • Liner ID: 0.75mm
  • Length: 74mm
  • OD: 6.3mm

Specifications

Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.