Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Westcott™ Straight Titanium Bonded Scissors
SCI_ED

Catalog No. S81747
Click to view available options
Grade:
Pre-K to 12

Provides three times the strength of classic stainless steel blades and they stay sharp and effective even after long use. The redesigned ergonomic handle provides maximum comfort during use.

  • Titanium Bonded blades are 3X harder than stainless steel; blades stay sharper longer
  • Contoured soft grip handles for maximum comfort
  • Suitable for right or left hand use
  • Corrosion resistant for longer blade life
  • Ideal for heavy usage cutting of plastic packages, thick fabric and boxes
  • Manufacturer's limited lifetime warranty

Specifications

Product Type Scissor
Color Gray and Yellow
Grade Pre-K to 12
Age 4 to 18
Quantity 2/Pk.
Length (English) 8 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.