Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596486 Supplier Invitrogen™ Supplier No. PA596486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAMRQTPLTC SGHTRPVVDL AFSGITPYGY FLISACKDGK PMLRQGDTGD WIGTFLGHKG AVWGATLNKD ATKAATAAAD FTAKVWDAVS GDELMTLAHK HIVKTVDFTQ DSNYLLTGGQ DKLLRIYDLN KPEAEPKEIS GHTSGIKKAL WCSEDKQILS ADDKTVRLWD HATMTEVKSL NFNMSVSSME YIPEGEILVI TYGRSIAFHS AVSLDPIKSF EAPATINSAS LHPEKEFLVA GGEDFKLYKY DYNSGEELES YKGHFGPIHC VRFSPDGELY ASGSEDGTLR LWQTVVGKTY GLWKCVLPEE DSGELAKPKI GFPETTEEEL EEIASENSDC IFPSAPDVKA; Positive Samples: Jurkat, HeLa, SW480, A-549, Mouse brain, Mouse lung, Mouse testis; Cellular Location: Cytoplasm, Nucleus.

STRAP is a 350 amino acid MAP activator with WD-40 repeats that is localized both in the cytoplasm and the nucleus. It is a ubiquitously expressed protein that acts as an inhibitor of TGF-beta signaling and an important regulator of cell proliferation. Stable expression of STRAP results in activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway and in down-regulation of the cyclin-dependent kinase inhibitor p21 (Cip1), which results in retinoblastoma protein hyperphosphorylation. Upregulation of STRAP might play a role in tumor development as an early event for colorectal cancers. It is the first component of the U snRNP assembly machinery that associates with SMN complex in a compartment-specific way and plays a crucial role in the intracellular distribution of SMN1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRAP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Strap
Gene Accession No. Q5XIG8, Q9Y3F4, Q9Z1Z2
Gene Alias AW557906; C78091; C79202; MAP activator with WD repeats; MAWD; PT-WD; serine/threonine kinase receptor associated protein; Serine-threonine kinase receptor-associated protein; STRAP; UNR-interacting protein; UNRIP; WD-40 repeat protein PT-WD
Gene Symbols Strap
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human STRAP (NP_0091093).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11171, 20901, 297699
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.