Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRAP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568529
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8529 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8529 Supplier Bethyl Laboratories, Inc Supplier No. A304735AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

STRAP is a 350 amino acid MAP activator with WD-40 repeats that is localized both in the cytoplasm and the nucleus. It is a ubiquitously expressed protein that acts as an inhibitor of TGF-beta signaling and an important regulator of cell proliferation. Stable expression of STRAP results in activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway and in down-regulation of the cyclin-dependent kinase inhibitor p21 (Cip1), which results in retinoblastoma protein hyperphosphorylation. Upregulation of STRAP might play a role in tumor development as an early event for colorectal cancers. It is the first component of the U snRNP assembly machinery that associates with SMN complex in a compartment-specific way and plays a crucial role in the intracellular distribution of SMN1.

Specifications

Antigen STRAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene STRAP
Gene Accession No. Q9Y3F4, Q9Z1Z2
Gene Alias MAWD, PT-WD, UNRIP, MAP activator with WD repeats, WD-40 repeat protein PT-WD, serine-threonine kinase receptor-associated protein, unr-interacting protein
Gene Symbols STRAP
Host Species Rabbit
Immunogen Between 300 and 350
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11171, 20901
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.