Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Straws & Connectors™
SDPSCI_ED

Catalog No. S13217
Click to view available options
Includes:
232 Straws, 168 Connectors, Guide
390 Straws, 315 Connectors, Guide

Make playhouses and forts, design buildings, and engineer space crafts with these straw and connectors!

  • Durable plastic straws fit snuggly on the connectors, but are flexible so you can build arches and tunnels
  • Appropriate for a wide age range and easy to clean
  • Age: 4+
  • Made in USA

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Includes 232 Straws, 168 Connectors, Guide
Quantity 400 Pieces
Languages English, Spanish, French, German
Subject STEM, Physics - Matter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.