Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scientific Cell Company Liquid Filter for Testing Photmetric Accuracy
Shop All Scientific Cell Company Inc. Products
Click to view available options
Type:
Liquid Filter
Description
- Certified at wavelengths: 235, 257, 313, 350nm
- Slit width: 2nm
- Supplied in high-quality storage case
Specifications
Specifications
Type | Liquid Filter |
For Use With (Application) | Testing photometric accuracy (1.25 Abs) in the UV range |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction