Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531739 Supplier Invitrogen™ Supplier No. PA531739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, MCF-7. Predicted reactivity: Mouse (98%), Rat (97%), Chicken (85%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in spermatogenesis and sperm function. Plays a role in regulation of cell growth. Binds to double-stranded DNA and RNA. Binds most efficiently to poly(I:C) RNA than to poly(dI:dC) DNA. Binds also to single-stranded poly(G) RNA. Binds non-specifically to the mRNA PRM1 3'-UTR and adenovirus VA RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRBP
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene STRBP
Gene Accession No. Q96SI9
Gene Alias 6430510M02Rik; 74 kDa double-stranded RNA-binding protein; AI852513; C230082I21Rik; C86322; DKFZp434N214; double-stranded RNA-binding protein p74; epididymis luminal protein 162; FLJ11307; FLJ14223; FLJ14984; HEL162; ILF3L; MGC21529; MGC3405; p74; spermatid perinuclear RNA binding protein; spermatid perinuclear RNA-binding protein; SPNR; Strbp
Gene Symbols STRBP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human STRBP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.