Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583896 Supplier Invitrogen™ Supplier No. PA583896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPTRVRGSLR ACIWAELQRR MAMPEPEWTT VGPELNGLDS KLLLDLPIQL MDRLSNESIM LVVELVQRAP EQLLALTPLH QAALAERALQ NLAPKETPVS GEVLETLGPL VGFLG Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 87%.

This gene encodes a protein that is associated with the hair bundle of the sensory hair cells in the inner ear. The hair bundle is composed of stiff microvilli called stereocilia and is involved with mechanoreception of sound waves. This gene is part of a tandem duplication on chromosome 15; the second copy is a pseudogene. Mutations in this gene cause autosomal recessive non-syndromic deafness. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STRC
Gene Accession No. Q7RTU9
Gene Alias DFNB16; Stereocilin; STRC
Gene Symbols STRC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STRC. Recombinant protein control fragment (Product #RP-108380).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.