Learn More
Science First™ Stream Table
SDPSCI_ED
Catalog No. s06669
Click to view available options
Material:
Plastic
With an integrated design, setup is a breeze — simply add water, turn on the pump and teach.
Stream tables are an essential part of the earth sciences class for the study of erosion, sediment deposition and the behavior of rivers and lakes. Use them to unfold these processes inside the classroom at an accelerated rate — easier than observing a real river over the course of decades.
  • Rugged plastic construction
  • Features five water jets
  • One water jet yields one piddly stream, while many jets form tributaries that form rivers
  • Integrated reservoir with sediment trap prevents particulates from clogging the pump
  • Clear acrylic sides allow views of sediment layers
  • Integrated prop holds the table at several angles
  • Removable dam creates a lake to illustrate how sediments form deltas
  • Fully watertight
  • Folds for storage
Order Info
Dimensions: 48 x 20 in. (122 x 50.8cm).
Specifications
Product Type Stream Table
Material Plastic
For Use With (Application) For the study of erosion, sediment deposition and the behavior of rivers and lakes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.