Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 4AA ProPolymer™ Lux Flashlight

Catalog No. p-3891875
Click to view available options
Battery Run Time:
155 hr.
6 hr.
Weight (English):
6.3 oz.
7.16 oz.
Lumen:
100 lm
67 lm

Includes

Wrist lanyard

The 4AA ProPolymer Lux is a durable polymer flashlight that provides a longer reaching beam and is ergonomically shaped for maximum hand comfort.

High performance 4AA flashlight features a body constructed of a polymer engineering resin that is virtually indestructible, shock resistant, non-conductive and lightweight.

  • One C4™ LED with a 50,000 hour lifetime
  • Deep reflector produces long-range brilliance and smooth beam
  • 4200 candela, 100 lumens
  • Push-button tail switch for one-handed operation
  • Impact-resistant polymer body
  • Unbreakable polycarbonate lens with scratch resistant coating
  • O-ring sealed, water resistant
  • Corrosion proof and non-conductive
  • Powered by four AA alkaline batteries, included
  • Four hours at full intensity, two additional hours of declining usable light
  • Power regulated for maximum output throughout battery life

Certifications

UL listed for Class I, Division 1; Class II, Division 1; and Class III hazardous locations

Specifications

Light Source C4 LED
Weight (English) 7.16 oz.
Weight (Metric) 0.216 kg
Battery Run Time 6 hr.
Candle Power 4200 cd
Color Yellow or Black
For Use With (Application) For maximum hand comfort
Includes Wrist lanyard connection
Lumen 100 lm
Material Polymer
Product Type Flashlight
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.