Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CD-Chex™ CD34
SDP

Catalog No. 23285520 Shop All Streck Laboratories Products
Click to view available options
Level:
1
1, 2
2
3
Quantity:
2 x 1 mL
4 x 1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CD-Chex CD34 is a three-level process control for enumerating CD34 positive cells by flow cytometry. It requires no dilution and provides reference values similar to levels found in bone marrow, cord blood and mobilized patient samples.

  • Only commercially available CD34 control with three levels.
  • No dilution required.
  • Whole blood control- resembles patient sample.
  • Compatible with flow cytometers using the ISHAGE protocol and enumeration kits.
  • 30-day open-vial stability, 90-day closed-vial stability.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

For Use With (Equipment) Flow cytometer
Type CD34 Flow Cytometry Control
Intended Use Used as a complete process control when evaluating CD34 positive cells
Level 2
Quantity 2 x 1 mL
Stability 30 Days Open, 90 Closed Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.