Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck CD-Chex™ Plus Immunophenotyping Control

Click to view available options
Level:
CD4 Low
CD4 Low, Normal
Normal
Quantity:
2 x 3 mL
4 x 3 mL
5 x 3 mL
10 x 3 mL
Format:
Cap-pierceable Plastic Vial
Description
- Assayed for 30 parameters, including T-lymphocytes, B-lymphocytes and NK cells.
- Contains a CD34+ population and is available in two clinically relevant CD4+ levels.
- Compatible with BD Biosciences and Beckman Coulter flow cytometry systems.
- 30-day open-vial stability, 90-day closed-vial stability.
- Manufacturer established assay ranges – no need to verify range per CAP requirements.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Specifications
Certifications/Compliance | CE |
Content And Storage | 2°C to 10°C |
Format | Cap-pierceable Plastic Vial |
For Use With (Equipment) | Beckman Coulter™ flow cytometry system |
Type | CD-Chex™ plus Immunophenotyping Control |
Intended Use | Immunophenotyping including RBC lysis, Antibody staining, instrument performance and operator technique |
Level | CD4 Low |
Quantity | 5 x 3 mL |
Source | Human Peripheral Blood Leukocytes and Erythrocytes |
Stability | 3 Months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction