Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Cell-Chex™ Cell Body Fluid Control
SDP

Catalog No. p-109315
Click to view available options
Level:
1- UC
1- UC, 2
1- UC/CC, 2
1-CC
2
Quantity:
2 x 2 mL
4 x 2 mL
6 x 2 mL
12 x 2 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Cell-Chex is a multi-level manual cerebrospinal and body fluid control for evaluating the accuracy and precision of hemacytometer RBC and WBC counts, urate and CPPD crystal identification and manual WBC differential procedures.

  • Assayed for the 5-part WBC differential.
  • Contains human white blood cells and CPPD crystals.
  • 30-day open-vial stability, 6-month closed-vial stability.
  • Validates the entire cell qualification and quantification procedure.
  • Long shelf life reduces the number of shipments and associated costs.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

Content And Storage 2°C to 10°C
For Use With (Equipment) A hemacytometer or WBC differential counts using a cytocentrifuge smear
Type Cell Body Fluid Control
Intended Use Body fluid procedural control for RBC and WBC counts, crystal identification and WBC differentiation
Level 1- UC, 2
Quantity 4 x 2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.