Learn More
Streck ESR-10 Manual Rack for the Modified Westergren Sed Rate
SDP
Catalog No. 11716350 Shop All Streck Laboratories Products
Change view
Click to view available options
Positions:
10
Catalog No. Positions
11-716-350 10
1 options
Catalog No. 11-716-350 Supplier Streck Supplier No. 240321
The ESR-10 Manual Rack is a manual Modified Westergren system that can determine the ESR of whole blood in 30 minutes using 1.2 mL Streck ESR-Vacuum Tubes.
  • Provides manual ESR results in 30 minutes.
  • ˃96% correlation to the Modified Westergren Method gives true sed-rate results.
  • Provides results in half the time compared to competitor methods.
  • Requires only 1.2 mL blood.
  • ESR-Vacuum Tubes offer safe direct-draw blood collection and preserve the integrity of the patient sample for up to 72 hours (2°C to 10°C).
Specifications
Product Type Sed Rate Tube Rack
Positions 10
For Use With (Equipment) 1.2 mL Streck ESR-vacuum Tubes