Streck Para 12™ Plus Whole Blood Hematology Control

Hematology control for the Abbott CELL-DYN™ 3200, 3500 and 3700 instruments

Manufacturer: Streck 218934

 View more versions of this product

Catalog No. 11-716-208

  Check Availability
Add to Cart

    Specifications

    Product Type Para 12™ plus whole blood control
    Format Vial
    Stability 75 Days
    For Use With (Application) For evaluating the accuracy and precision of hematology instruments that provide a WBC differential
    For Use With (Equipment) Abbott CELL-DYN™ 3200, 3500, and 3700
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 4 x 3mL,4 x 3mL
    Source Stabilized human or mammalian RBC, mammalian or simulated white blood cells and a platelet component,Stabilized human or mammalian RBC, mammalian or simulated white blood cells and a platelet component
    Level 4 High,4 High

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit