Learn More
Streck Retic-Chex™ Linearity
SDP
Catalog No. p-109400
Click to view available options
Quantity:
5 x 1 mL
5 x 3 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Retic-Chex Linearity for BC is designed to establish the reportable range and linearity of Beckman Coulter™ hematology analyzers that report the reticulocyte parameter.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • Assayed for Beckman Coulter hematology analyzers.
  • 5-day open-vial stability, 105-day closed-vial stability.
  • Established assay ranges provide less room for error.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Compliance
Complies with CLIA 88 Regulations [42 CFR 493.1217 (b)(2)(ii)(C)], and CAP Sections 02.2576 and 02.2579 for calibration verification.
Specifications
Certifications/Compliance CLIA-88
Type Linearity
Intended Use Calibration verification assessment, establishing the analytic linearity of automated Reticulocyte counting instruments
Level 1 to 5
Quantity 5 x 3 mL
Stability 8 Days Open Vial
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.