Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Sickle-Chex™ Positive and Negative Controls
Compatible with both commercial sickle screening tests and hemoglobin electrophoresis
Supplier: Streck 217653
This item is not returnable.
View return policy
Description
- Packaged in 2.5mL squeeze dropper vials
- No pipetting is required
- Whole blood control
- Positive and negative levels
- 180-day closed-vial stability
- 100-day open-vial stability
- Streck SICKLEDEX™
- Siemens™ Sickle-Sol™ Test
- Pacific Hemostasis™ SickleScreen™ Sickling Hemoglobin Screening Kit
Order Info
Packaged in 2.5ml squeeze dropper vials for easy sample dispensing without pipeting; does not require reconstitution prior to use, so it can be used like a patient sample
Specifications
2°C to 10°C | |
Sickle-Chex™ Positive and Negative Controls | |
2 x 2.5 mL |
Safety and Handling
ShelfLife : 6 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction