Learn More
Streck Sickledex™ Solubility Kit
Shop All Streck Laboratories Products
Click to view available options
Quantity:
100 Tests
12 Tests
50 Tests
Description
Includes
Each test kit contains two bottles of phosphate buffer with Saponin, two buffer dispensing caps and two vials of Sodium Hydrosulfite reagent powder.
Requires
Each test requires 2 mL of reconstituted solubility buffer and 20μL of anticoagulated whole blood or 20μL of sickle cell hemoglobin control.
- Test results available in 6 minutes.
- Requires only 20 µL of patient or control sample.
- 45-day open-vial stability, 12-month closed-vial stability.
- Buffer packaging reduces waste and maximizes open-vial stability.
- No pipetting necessary.
- Sickle-Chex whole blood control can be used like a patient sample.
Order Info
Additional test tube racks are available separately, Cat. No. 11-716-452. Additional test tubes, Cat. No. NC1459622.
Specifications
Specifications
Type | Sickle Cell Solubility Test Kit |
Includes | 2 Bottles of Phosphate Buffer with Saponin, 2 Buffer Dispensing Caps, 2 Vials of Sodium Hydrosulfite Reagent Powder |
Quantity | 50 Tests |
Test Time | 6 min. |