Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck STaK-Chex™ Plus Retics Control

Click to view available options
Level:
1 Low, 1 Normal, 1 High
12 Normal
4 High
4 Low
4 Low, 4 Normal, 4 High
Quantity:
3 x 4.5 mL
4 x 4.5 mL
12 x 4.5 mL
Format:
Piercable-cap Vial
Vial
Description
- Includes all CBC parameters, reticulocytes and nucleated RBCs.
- Available in three clinically significant levels.
- Plastic vials with pierceable caps for analyzer autosampling.
- Assay values are available for download.
- 14-day open-vial stability, 105-day closed-vial stability.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Specifications
Certifications/Compliance | CE |
Content And Storage | 2°C to 10°C |
Format | Vial |
For Use With (Equipment) | Beckman Coulter™ UniCel™ DxH™ 800, LH780/785, LH 750/755, LH500 and HmX |
Type | STaK-Chex™ plus Retics Control |
Includes | Assay Disc |
Intended Use | For evaluating the accuracy and precision of automated procedures that measure Blood cell parameters |
Level | 4 Low, 4 Normal, 4 High |
Quantity | 12 x 4.5 mL |
Stability | 105 Days |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction