Learn More
Description
- Individually serial-numbered and NIST-traceable.
- Yellow background for easier reading.
- Enclosed in a durable plastic case to contain contents in case of breakage.
- A wide temperature range allows Temp-Chex thermometers to be used to monitor most laboratory applications.
- Mercury free and non-hazardous for increased laboratory safety.
- Thermometer probe is housed in a vial of propylene glycol, which creates a temperature buffer that prevents fluctuations.
Certifications
NIST, DKD Traceable: Each thermometer engraved with serial number traceable to NIST/DKD.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | NIST, DKD |
| For Use With (Application) | Refrigerator |
| Includes | Temperature wall chart |
| Thermometer Fill | Red Spirit |
| Product Type | Thermometer |
| Max. Temperature (Metric) | 15°C |
| Min. Temperature (Metric) | -5°C |
| Temperature Range (Metric) | -5°C to +15°C |
| Accuracy | ±0.5°C |
| Subdivision | 0.5° |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

