Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Temp-Chex™ Thermometer
GSA_VASDP

Catalog No. 11716186C Shop All Streck Laboratories Products
Change view
Click to view available options
Temperature Range (Metric):
-30°C to +1°C
-5°C to +15°C
-90°C to +25°C
18°C to 50°C
For Use With (Application):
Freezers
Incubator
Refrigerator
Ultralow freezer
Thermometer Fill:
Red Spirit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric) For Use With (Application) Thermometer Fill
11-716-186C -90°C to +25°C Ultralow freezer Red Spirit
11-716-186B -30°C to +1°C Freezers Red Spirit
11-716-187B 18°C to 50°C Incubator Red Spirit
11-716-185B -5°C to +15°C Refrigerator Red Spirit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-716-186C Supplier Streck Supplier No. 240047
This item is not returnable. View return policy

Temp-Chex with Red Spirit is a certified NIST-traceable laboratory thermometer for monitoring refrigerators, ultra-low freezers, incubators, and room temperatures.

  • Individually serial-numbered and NIST-traceable.
  • Yellow background for easier reading.
  • Enclosed in a durable plastic case to contain contents in case of breakage.
  • A wide temperature range allows Temp-Chex thermometers to be used to monitor most laboratory applications.
  • Mercury free and non-hazardous for increased laboratory safety.
  • Thermometer probe is housed in a vial of propylene glycol, which creates a temperature buffer that prevents fluctuations.

Certifications

NIST, DKD Traceable: Each thermometer engraved with serial number traceable to NIST/DKD.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance NIST, DKD
For Use With (Application) Ultralow freezer
Includes Temperature wall chart
Thermometer Fill Red Spirit
Product Type Thermometer
Max. Temperature (Metric) 25°C
Min. Temperature (Metric) -90°C
Temperature Range (Metric) -90°C to +25°C
Accuracy ±0.5°C
Subdivision 0.5°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.