Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strep II Tag Antibody (517), Novus Biologicals™
SDP

Catalog No. NBP243735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243735 Supplier Novus Biologicals Supplier No. NBP243735
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence

Strep II Tag Monoclonal antibody specifically detects Strep II Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen Strep II Tag
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 517
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS, 20% Glycerol (pH7) with No Preservative
Host Species Mouse
Immunogen Immunogen Amino acid sequence - ASWSHPQFEKGA
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.