Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Strep Tag II Monoclonal Antibody (A5D10)
GREENER_CHOICE

Catalog No. PIMA544988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544988 Supplier Invitrogen™ Supplier No. MA544988
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Strep Tag II Monoclonal Antibody for Western Blot

The Strep-tag II is an eight-residue minimal peptide sequence (Trp-Ser-His-Pro-Gln-Phe-Glu-Lys) that exhibits intrinsic affinity toward streptavidin and can be fused to recombinant proteins in various fashions. The original Strep-tag was a nine amino acid peptide with high specificity and affinity towards streptavidin which allows the simple purification of protein by use of affinity columns, but required addition to only the C-terminus of recombinant proteins. To also allow a Strep-tag to be placed at the N-terminus of recombinant proteins, it was re-engineered and re-named Strep-tag II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Strep Tag II
Applications Western Blot
Classification Monoclonal
Clone A5D10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias Streptavidin; Streptomyces
Host Species Mouse
Immunogen Synthetic peptide corresponding to Strep-tag II.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.