Learn More
Invitrogen™ Strep Tag II Monoclonal Antibody (A5D11)

Description
Positive Control: Strep-Tag II protein transfected 293F cells lysate, Non-transfected 293F cells lysate.
Specifications
Specifications
| Antigen | Strep Tag II |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A5D11 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene Alias | Streptavidin; Streptomyces |
| Host Species | Mouse |
| Immunogen | Synthetic peptide corresponding to Strep-tag II. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.