Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Streptavidin, Alexa Fluor™ 594 conjugate Non-distribution product as customer accommodation.

Streptavidin, Alexa Fluor™ 594 conjugate

$245.00 - $287.00

Specifications

Binding Capacity Very high binding affinity for biotin
Color Purple
Detection Method Fluorescent
For Use With (Application) Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more
Packaging 1 Each
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S11227A Non-distribution product as customer accommodation. View Documents Molecular Probes™
S11227
1mg Each for $245.00
Add to cart
 
S32356A Non-distribution product as customer accommodation. View Documents Molecular Probes™
S32356
0.5mL Each for $287.00
Add to cart
 
Description

Description

Alexa Fluor™ 594 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 594 Streptavidin Conjugates:
  • Alexa Fluor™ 594 streptavidin conjugate has Ex/Em maxima of ~ (590/617)
  • Bright, photostable fluorescence
  • High solubility in aqueous solutions
  • Available in multiple colors
  • Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

    Properties of Alexa Fluor™ Dyes
    Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

    Blocking Endogenous Biotin
    Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

    Related Links:

    Learn more about Avidin-Biotin Detection

    Learn more about Alexa Fluor™ Dyes

    Find out about other Labeled Streptavidin Conjugates

    Read Avidin and Streptavidin Conjugates-Section 7.6 in the Molecular Probes Handbook
  • Specifications

    Specifications

    Very high binding affinity for biotin
    Fluorescent
    1 Each
    TLC
    2mg/mL
    Lyophilized
    Purple
    Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more
    Alexa Fluor™ 594 Conjugate Streptavidin
    For research use only
    Alexa Fluor™ 594
    Alexa Fluor™ 594 Conjugate Streptavidin
    Safety and Handling

    Safety and Handling

    ShelfLife : 6 Month(s)

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit