Learn More
R&D Systems™ Streptavidin Check&Go! Control Kit
SDP
Catalog No. 40040030 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
40040030 30 Tests
1 options
This product is for research use only and is not approved for use in humans or in clinical diagnosis
Catalog No. 40040030 Supplier R&D Systems™ Supplier No. 40040030
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This product is for research use only and is not approved for use in humans or in clinical diagnosis
A Novus product, part of R&D Systems. Control kit
  • Components:
    • 30 Protein A/G strips
    • 2 Cryovials of 10x Running Buffer
    • 1 Vial of 40nm InnovaCoat Gold Biotin
    • 1 Vial of Positive Control
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis
  • Kits are guaranteed for 6 months from date of receipt
Specifications
Product Line Novus
Kit Contents 30 protein A/G strips, 1 vial of 40 nm InnovaCoat Gold biotin, 1 vial of positive control, 2 cryovials of 10X running buffer
Quantity 30 Tests