Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Streptavidin-CY5 (ZyMax Grade)

Catalog No. 438316
Encompass_Preferred
Click to view available options
Quantity:
1 mL

Streptavidin-CY5 (ZyMax Grade)

Applications: Western blotting, ELISA, Immunohistochemistry, ISH

ZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Conjugate Cy5
Quantity 1 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Line Invitrogen™
Product Type Streptavidin Conjugate (fluorescent)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.