Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Streptavidin Native Protein
SDP

Catalog No. NBP229370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229370 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229370 Supplier Novus Biologicals™ Supplier No. NBP229370
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native Streptavidin. (Accession #: P22629) The Streptavidin Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Specifications

Formulation Lyophilized: 0.02 M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 0.01% (w/v) Gentamicin Sulfate. 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free.
Quantity 1 mg
Content And Storage 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
For Use With (Application) Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purity or Quality Grade Chromatography
Product Type Streptavidin Protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.