Learn More
Description
Specifications
Specifications
| Formulation | 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free; 0.05% Sodium Azide |
| Quantity | 1 mg |
| Content And Storage | 4°C. Do not freeze. |
| For Use With (Application) | WB, ELISA, IHC |
| Product Type | Streptavidin Protein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
