Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptavidin Polyclonal Antibody, Biotin, Rockland™
SDP

Catalog No. 501990979
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0979 25 μL
50-199-0978 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0979 Supplier Rockland Immunochemicals Supplier No. 200406095S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Rabbit Serum and Streptavidin. No reaction was observed against Avidin.

Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ∼ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.

Specifications

Antigen Streptavidin
Applications Dot Blot, ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Streptavidin (Streptomyces avidinii)
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.