Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococci Group B Monoclonal Antibody (HSB11-238.6), QED Bioscience™
SDP

Catalog No. 502277611
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7611 1 mg
50-227-7612 100 μg
50-227-7613 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7611 Supplier Leinco Technologies Inc Supplier No. 156041000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Streptococci Group B Monoclonal antibody specifically detects Streptococci Group B in Bacteria samples. It is validated for ELISA

Specifications

Antigen Streptococci Group B
Applications ELISA
Classification Monoclonal
Clone HSB11-238.6
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen Pool of UV-inactivated Group B Streptococci Types Ia, Ib, Ic, II, and III
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.