Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococci Group B Monoclonal Antibody (HSB12-396.1), QED Bioscience™

Mouse Monoclonal Antibody
$250.22 - $1002.00
Specifications
Antigen | Streptococci Group B |
---|---|
Clone | HSB12-396.1 |
Concentration | 0.5-1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7615
![]() ![]() |
Leinco Technologies Inc
15605100UG |
100 μg |
Each for $250.22
|
|
|||||
50-227-7616
![]() ![]() |
Leinco Technologies Inc
15605500UG |
500 μg |
Each for $500.43
|
|
|||||
50-227-7614
![]() ![]() |
Leinco Technologies Inc
156051000UG |
1 mg |
Each for $1,002.00
|
|
|||||
Description
Streptococci Group B Monoclonal antibody specifically detects Streptococci Group B in Bacteria samples. It is validated for ELISASpecifications
Streptococci Group B | |
0.5-1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
IgG2a | |
Protein A | |
Antibody |
HSB12-396.1 | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
Pool of UV-inactivated Group B Streptococci Types Ia, Ib, Ic, II, and III | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title