Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus Agalactiae Antibody (224/46), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062933AF647
Description
Streptococcus Agalactiae Monoclonal specifically detects Streptococcus Agalactiae in Bacteria samples. It is validated for ELISA.Specifications
Streptococcus Agalactiae | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
This product is specific for Streptococcus agalactiae a gram-positive streptococcus characterized by the presence of the group B Lancefield antigen. | |
Store at 4C in the dark. | |
IgG3 |
ELISA | |
224/46 | |
ELISA | |
s agalactiae, S. agalactiae | |
Streptococcus agalactiae (NCTC 11360) | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction